Search
Search
| Gene Symbol: NR1I3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037996.1 | NP_001033085.1 | 4 - 5 | 625 | 68 | |
| XM_013996822.2 | XP_013852276.1 | 903 | 68 | |||
| GenBank mRNA | AB214979.1 | - | 4 - 5 | 625 | 68 | |
| AK232831.1 | - | 4 - 5 | 625 | 68 | ||
| AK346033.1 | - | 4 - 5 | 625 | 68 | ||
| DQ249306.1 | - | 3 - 4 | 415 | 68 | ||
| KF586862.1 | - | 3 - 4 | 409 | 68 | ||
| KF586863.1 | - | 3 - 4 | 409 | 68 | ||
| KF586864.1 | - | 3 - 4 | 409 | 68 | ||
Set Membership: |
|