Search
Search
| Gene Symbol: APOM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040640.1 | NP_001035730.1 | 4 - 5 | 491 | 77 | |
| GenBank mRNA | AY610077.1 | - | 4 - 5 | 524 | 77 | |
| DQ329240.2 | - | 4 - 5 | 491 | 77 | ||
Set Membership: |
|