Search
Search
| Gene Symbol: SAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213814.2 | NP_998979.1 | 1 - 2 | 164 | 74 | |
| GenBank mRNA | AJ249974.1 | - | 1 - 2 | 164 | 74 | |
| AK350053.1 | - | 1 - 2 | 74 | |||
| AK399206.1 | - | 1 - 2 | 74 | |||
Set Membership: |
|