Search
Search
| Gene Symbol: MYH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213855.2 | NP_999020.2 | 25 - 26 | 3362 | 72 | |
| GenBank mRNA | AB053226.1 | - | 23 - 24 | 3268 | 72 | |
| U75316.1 | - | 24 - 25 | 3323 | 72 | ||
Set Membership: |
|