Search
Search
| Gene Symbol: FBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213979.1 | NP_999144.1 | 7 | 1056 | 96 | |
| GenBank mRNA | AK233435.1 | - | 7 | 1087 | 96 | |
| AK391160.1 | - | 7 | 1088 | 96 | ||
| M86347.1 | - | 7 | 1056 | 96 | ||
Set Membership: |
|