Search
Search
| Gene Symbol: DGAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214051.1 | NP_999216.1 | 17 | 1551 | 72 | |
| XM_005655311.3 | XP_005655368.2 | 1498 | 72 | |||
| XM_021088697.1 | XP_020944356.1 | 1646 | 72 | |||
| GenBank mRNA | AY093657.1 | - | 17 | 1551 | 72 | |
Set Membership: |
|