Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Ss03393808_u1
          See other MT3 GE Assays ›
          Gene Symbol:
          MT3
          Gene Name:
          metallothionein 3
          Gene Aliases:
          MT-III
          Chromosome Location:
          Chr.6: 18634735 - 18636134 on Build Sscrofa11.1
          Species:
          Pig
          Species Specific ID (Flybase ID):
          -
          Assay ID Ss03393808_u1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MT3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_214056.1 NP_999221.1 3 148 83
          GenBank mRNA AK392719.1 - 4 241 147
          U95969.1 - 3 148 83

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          positive regulation of protein phosphorylation
          energy reserve metabolic process
          cholesterol catabolic process
          zinc II ion transport
          cellular zinc ion homeostasis
          apoptotic process
          negative regulation of autophagy
          positive regulation of gene expression
          positive regulation of necrotic cell death
          positive regulation of cell death
          astrocyte development
          histone modification
          removal of superoxide radicals
          negative regulation of cell growth
          negative regulation of axon extension
          positive regulation of vascular endothelial growth factor receptor signaling pathway
          regulation of response to food
          activation of protein kinase B activity
          leptin-mediated signaling pathway
          cellular response to oxidative stress
          cellular response to drug
          positive regulation of transcription from RNA polymerase II promoter in response to oxidative stress
          negative regulation of apoptotic process
          positive regulation of catalytic activity
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          protein kinase B signaling
          negative regulation of neuron apoptotic process
          cellular lipid catabolic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          protein stabilization
          negative regulation of oxidoreductase activity
          zinc ion homeostasis
          cadmium ion homeostasis
          regulation of protein glycosylation
          negative regulation of necrotic cell death
          ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          cellular response to cadmium ion
          cellular response to nitric oxide
          positive regulation of lysosomal membrane permeability
          negative regulation of cysteine-type endopeptidase activity
          positive regulation of oxygen metabolic process
          negative regulation of reactive oxygen species metabolic process
          copper ion binding
          drug binding
          zinc ion binding
          protein kinase activator activity
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process
          cadmium ion binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline