Search
Search
| Gene Symbol: SFTPD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214110.1 | NP_999275.1 | 6 - 7 | 711 | 60 | |
| XM_005671145.3 | XP_005671202.1 | 929 | 60 | |||
| XM_013983376.2 | XP_013838830.1 | 758 | 60 | |||
| GenBank mRNA | AF132496.2 | - | 6 - 7 | 711 | 60 | |
Set Membership: |
|