Search
Search

AMY2
LOC100521789| Gene Symbol: AMY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214195.1 | NP_999360.1 | 4 - 5 | 741 | 58 | |
| XM_005663621.3 | XP_005663678.2 | 964 | 58 | |||
| XM_005663623.3 | XP_005663680.2 | 1050 | 58 | |||
| XM_021088708.1 | XP_020944367.1 | 1046 | 58 | |||
| XM_021088709.1 | XP_020944368.1 | 1170 | 58 | |||
| XM_021088710.1 | XP_020944369.1 | 863 | 58 | |||
| XM_021088711.1 | XP_020944370.1 | 806 | 58 | |||
| GenBank mRNA | AF064742.1 | - | 4 - 5 | 741 | 58 | |
| AK395394.1 | - | 5 - 6 | 902 | 58 | ||
| Gene Symbol: LOC100521789 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125881.6 | XP_003125929.2 | 1092 | 58 | ||
| XM_005663625.3 | XP_005663682.2 | 924 | 58 | |||
| XM_021090122.1 | XP_020945781.1 | 1030 | 58 | |||
| XM_021090123.1 | XP_020945782.1 | 1080 | 58 | |||
| XM_021090124.1 | XP_020945783.1 | 806 | 58 | |||
Set Membership: |
|