Search
Search
| Gene Symbol: OPALIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214210.1 | NP_999375.1 | 5 - 6 | 832 | 110 | |
| XM_013983587.2 | XP_013839041.1 | 965 | 110 | |||
| XM_013983588.2 | XP_013839042.1 | 869 | 110 | |||
| XM_013983589.2 | XP_013839043.1 | 1215 | 110 | |||
| XM_013983590.2 | XP_013839044.1 | 683 | 110 | |||
| XM_013983591.2 | XP_013839045.1 | 600 | 110 | |||
| XM_013983592.2 | XP_013839046.1 | 1105 | 110 | |||
| XM_013983593.2 | XP_013839047.1 | 573 | 110 | |||
| XM_013983594.2 | XP_013839048.1 | 104 | 110 | |||
| XM_021072121.1 | XP_020927780.1 | 727 | 110 | |||
| GenBank mRNA | AK392173.1 | - | 5 - 6 | 570 | 110 | |
| AK400134.1 | - | 5 - 6 | 632 | 110 | ||
| AK400143.1 | - | 5 - 6 | 642 | 110 | ||
| AK400203.1 | - | 5 - 6 | 496 | 110 | ||
| AK400205.1 | - | 4 - 5 | 260 | 110 | ||
| X89480.1 | - | 832 | 110 | |||
| Y11026.1 | - | 552 | 110 | |||
Set Membership: |
|