Search
Search
| Gene Symbol: HYAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214440.1 | NP_999605.1 | 2 - 3 | 1027 | 63 | |
| XM_005669574.3 | XP_005669631.1 | 1217 | 63 | |||
| XM_005669575.3 | XP_005669632.1 | 2301 | 63 | |||
| XM_005669576.3 | XP_005669633.1 | 2764 | 63 | |||
| XM_013981684.1 | XP_013837138.1 | 1523 | 63 | |||
| XM_021068571.1 | XP_020924230.1 | 1136 | 63 | |||
| GenBank mRNA | AK343526.1 | - | 3 - 4 | 1186 | 63 | |
| AY497544.1 | - | 2 - 3 | 1027 | 63 | ||
| EU925135.1 | - | 2 - 3 | 996 | 63 | ||
Set Membership: |
|