Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
我們的服務對象
生物技術
生物製藥
CDMO
實驗室診斷
工業與應用科學
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ss03816281_m1
See other TCF7L2 GE Assays ›
Gene Symbol:
TCF7L2
Gene Name:
transcription factor 7 like 2
Gene Aliases:
-
Chromosome Location:
Chr.14: 123343695 - 123547332 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID
Ss03816281_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: TCF7L2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_001928695.5
XP_001928730.1
1748
84
XM_003133187.4
XP_003133235.1
1817
84
XM_003133188.4
XP_003133236.1
1735
84
XM_003133189.5
XP_003133237.1
1889
84
XM_003359370.4
XP_003359418.1
1747
84
XM_003359371.4
XP_003359419.1
1747
84
XM_003359373.4
XP_003359421.1
1815
84
XM_003359374.4
XP_003359422.1
1746
84
XM_005657462.3
XP_005657519.1
1979
84
XM_005657463.3
XP_005657520.1
1976
84
XM_005657464.3
XP_005657521.1
1968
84
XM_005657465.3
XP_005657522.1
1963
84
XM_005657466.3
XP_005657523.1
1952
84
XM_005657467.3
XP_005657524.1
1979
84
XM_005657468.3
XP_005657525.1
1979
84
XM_005657469.3
XP_005657526.1
1910
84
XM_005657470.3
XP_005657527.1
1907
84
XM_005657471.3
XP_005657528.1
1833
84
XM_005657472.3
XP_005657529.1
1817
84
XM_005657473.3
XP_005657530.1
1833
84
XM_005657474.3
XP_005657531.1
1764
84
XM_005657475.3
XP_005657532.1
1753
84
XM_005657476.3
XP_005657533.1
1737
84
XM_005657477.3
XP_005657534.1
1764
84
XM_005657478.3
XP_005657535.1
1980
84
XM_005657482.3
XP_005657539.1
1816
84
XM_005657483.3
XP_005657540.1
1764
84
XM_005657484.3
XP_005657541.1
1832
84
XM_005657486.3
XP_005657543.1
1804
84
XM_005657487.3
XP_005657544.1
1746
84
XM_005657489.3
XP_005657546.1
1735
84
XM_013990492.2
XP_013845946.1
1979
84
XM_021073288.1
XP_020928947.1
1963
84
XM_021073290.1
XP_020928949.1
1963
84
XM_021073291.1
XP_020928950.1
1960
84
XM_021073292.1
XP_020928951.1
1894
84
XM_021073293.1
XP_020928952.1
1894
84
XM_021073294.1
XP_020928953.1
1806
84
XM_021073295.1
XP_020928954.1
1833
84
XM_021073296.1
XP_020928955.1
1817
84
XM_021073297.1
XP_020928956.1
1806
84
XM_021073299.1
XP_020928958.1
1736
84
XM_021073300.1
XP_020928959.1
1747
84
XM_021073301.1
XP_020928960.1
1963
84
XM_021073302.1
XP_020928961.1
1894
84
XM_021073303.1
XP_020928962.1
1816
84
XM_021073304.1
XP_020928963.1
1833
84
XM_021073305.1
XP_020928964.1
1816
84
XM_021073306.1
XP_020928965.1
1747
84
XM_021073307.1
XP_020928966.1
1736
84
GenBank mRNA
AK347963.1
-
10 - 11
1698
84
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
3' Most
Recommended Assay
Sus scrofa
Probe spans exons
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline