Search
Search
| Gene Symbol: OSTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098597.1 | NP_001092067.1 | 3 | 253 | 89 | |
| GenBank mRNA | EF369511.1 | - | 2 | 200 | 89 | |
| EF503638.1 | - | 3 | 253 | 89 | ||
| EU122441.1 | - | 2 | 197 | 89 | ||
Set Membership: |
|