Search
Search
| Gene Symbol: ASF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137625.1 | NP_001131097.1 | 3 - 4 | 495 | 59 | |
| GenBank mRNA | AK344388.1 | - | 3 - 4 | 526 | 59 | |
| FJ175659.1 | - | 3 - 4 | 495 | 59 | ||
Set Membership: |
|