Search
Search
| Gene Symbol: FITM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128470.1 | NP_001121942.1 | 1 - 2 | 266 | 72 | |
| GenBank mRNA | EU625275.1 | - | 1 - 2 | 266 | 72 | |
| FJ393218.1 | - | 1 - 2 | 660 | 72 | ||
Set Membership: |
|