Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144842.2 | NP_001138314.1 | 3 - 4 | 323 | 70 | |
GenBank mRNA | AK391822.1 | - | 3 - 4 | 351 | 70 | |
FJ487633.1 | - | 3 - 4 | 324 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |