Search
Search
| Gene Symbol: CLDN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160076.1 | NP_001153548.1 | 2 - 3 | 562 | 68 | |
| XM_005669183.3 | XP_005669240.1 | 508 | 68 | |||
| XM_005669184.2 | XP_005669241.1 | 667 | 68 | |||
| XM_021066034.1 | XP_020921693.1 | 669 | 68 | |||
| GenBank mRNA | AK239948.1 | - | 2 - 3 | 778 | 68 | |
| AK392970.1 | - | 2 - 3 | 785 | 68 | ||
| FJ887979.1 | - | 2 - 3 | 562 | 68 | ||
Set Membership: |
|