Search
Search
| Gene Symbol: LDHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172363.2 | NP_001165834.1 | 1 - 2 | 86 | 63 | |
| XM_013994501.2 | XP_013849955.2 | 736 | 63 | |||
| GenBank mRNA | AK236433.1 | - | 1 - 2 | 124 | 63 | |
| AK343612.1 | - | 1 - 2 | 77 | 63 | ||
| AK343811.1 | - | 1 - 2 | 75 | 63 | ||
| AK391205.1 | - | 1 - 2 | 77 | 63 | ||
| AK391208.1 | - | 1 - 2 | 74 | 63 | ||
| AK391657.1 | - | 1 - 2 | 78 | 63 | ||
| AK395255.1 | - | 1 - 2 | 110 | 63 | ||
| AK399447.1 | - | 1 - 2 | 110 | 63 | ||
| AK399466.1 | - | 1 - 2 | 77 | 63 | ||
| AK399472.1 | - | 1 - 2 | 93 | 63 | ||
| AK399544.1 | - | 1 - 2 | 77 | 63 | ||
| AK399555.1 | - | 1 - 2 | 77 | 63 | ||
| AK399560.1 | - | 3 - 4 | 286 | 63 | ||
| FJ865398.1 | - | 1 - 2 | 87 | 63 | ||
| U07178.1 | - | 1 - 2 | 73 | 63 | ||
Set Membership: |
|