Search
Search

SPATA24| Gene Symbol: SPATA24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177922.1 | NP_001171393.1 | 4 - 5 | 409 | 106 | |
| XM_005661705.3 | XP_005661762.1 | 381 | 106 | |||
| GenBank mRNA | AK392880.1 | - | 4 - 5 | 410 | 106 | |
| GU475005.1 | - | 4 - 5 | 409 | 106 | ||
Set Membership: |
|