Search
Search

DCSTAMP| Gene Symbol: DCSTAMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185150.1 | NP_001172079.1 | 3 - 4 | 1387 | 87 | |
| XM_021088629.1 | XP_020944288.1 | 2347 | 87 | |||
| GenBank mRNA | AK231004.1 | - | 3 - 4 | 1387 | 87 | |
| AK389845.1 | - | 3 - 4 | 1387 | 87 | ||
Set Membership: |
|