Search
Search
| Gene Symbol: PVALB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190157.1 | NP_001177086.1 | 2 - 3 | 199 | 80 | |
| XM_005663796.3 | XP_005663853.1 | 305 | 80 | |||
| XM_021090958.1 | XP_020946617.1 | 400 | 80 | |||
| XM_021090960.1 | XP_020946619.1 | 253 | 80 | |||
| GenBank mRNA | AK392706.1 | - | 2 - 3 | 223 | 80 | |
| AK400856.1 | - | 3 - 4 | 269 | 80 | ||
| AY609522.1 | - | 2 - 3 | 195 | 80 | ||
| BX915869.2 | - | 2 - 3 | 218 | 80 | ||
| BX919484.2 | - | 3 - 4 | 272 | 80 | ||
| FM994923.1 | - | 2 - 3 | 192 | 80 | ||
Set Membership: |
|