Search
Search
| Gene Symbol: MCCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021069887.1 | XP_020925546.1 | 928 | 70 | ||
| XM_021069888.1 | XP_020925547.1 | 626 | 70 | |||
| XM_021069889.1 | XP_020925548.1 | 574 | 70 | |||
| GenBank mRNA | AK240078.1 | - | 5 - 6 | 575 | 70 | |
| AK390589.1 | - | 5 - 6 | 599 | 70 | ||
Set Membership: |
|