Search
Search
| Gene Symbol: MOSPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195356.1 | NP_001182285.1 | 1 - 2 | 272 | 92 | |
| GenBank mRNA | AK240357.1 | - | 2 - 3 | 333 | 92 | |
| GU373666.1 | - | 1 - 2 | 272 | 92 | ||
Set Membership: |
|