Search
Search

FOXRED1| Gene Symbol: FOXRED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244921.1 | NP_001231850.1 | 1 - 2 | 112 | 88 | |
| XM_005667490.3 | XP_005667547.1 | 143 | 88 | |||
| GenBank mRNA | EW283212.2 | - | 1 - 2 | 112 | 88 | |
Set Membership: |
|