Search
Search

FOXRED1| Gene Symbol: FOXRED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244921.1 | NP_001231850.1 | 9 - 10 | 1128 | 90 | |
| XM_005667490.3 | XP_005667547.1 | 1159 | 90 | |||
| XM_013979554.2 | XP_013835008.1 | 1118 | 90 | |||
| GenBank mRNA | AK232917.1 | - | 7 - 8 | 750 | 90 | |
Set Membership: |
|