Search
Search
| Gene Symbol: RPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244942.1 | NP_001231871.1 | 3 - 4 | 327 | 83 | |
| GenBank mRNA | AK230885.1 | - | 3 - 4 | 313 | 83 | |
| AY609526.1 | - | 3 - 4 | 327 | 83 | ||
Set Membership: |
|