Search
Search
| Gene Symbol: TADA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206536.1 | NP_001193465.1 | 5 - 6 | 850 | 85 | |
| XM_021068275.1 | XP_020923934.1 | 825 | 85 | |||
| GenBank mRNA | AK233113.1 | - | 5 - 6 | 809 | 85 | |
| AK235953.1 | - | 5 - 6 | 850 | 85 | ||
| AK238770.1 | - | 5 - 6 | 755 | 85 | ||
| AK392946.1 | - | 5 - 6 | 978 | 85 | ||
Set Membership: |
|