Search
Search
| Gene Symbol: HEY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243329.1 | NP_001230258.1 | 2 - 3 | 299 | 88 | |
| XM_013992576.2 | XP_013848030.1 | 198 | 88 | |||
| XM_021086293.1 | XP_020941952.1 | 998 | 88 | |||
| GenBank mRNA | BP149699.1 | - | 2 - 3 | 300 | 88 | |
Set Membership: |
|