Search
Search
| Gene Symbol: PRKAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243621.1 | NP_001230550.1 | 3 - 4 | 794 | 72 | |
| XM_005670662.3 | XP_005670719.1 | 809 | 72 | |||
| GenBank mRNA | CN158460.1 | - | 2 - 3 | 213 | 72 | |
| DY407095.1 | - | 3 - 4 | 279 | 72 | ||
Set Membership: |
|