Search
Search
| Gene Symbol: EXOSC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243824.1 | NP_001230753.1 | 1 - 2 | 65 | 62 | |
| GenBank mRNA | AK395369.1 | - | 1 - 2 | 84 | 62 | |
| AY610143.1 | - | 1 - 2 | 65 | 62 | ||
Set Membership: |
|