Search
Search
| Gene Symbol: RPS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244070.1 | NP_001230999.1 | 2 - 3 | 422 | 135 | |
| XM_005664714.3 | XP_005664771.2 | 311 | 135 | |||
| GenBank mRNA | AK233375.1 | - | 2 - 3 | 169 | 135 | |
| AK236665.1 | - | 727 | 135 | |||
| AK347721.1 | - | 2 - 3 | 170 | 135 | ||
| AK392151.1 | - | 2 - 3 | 227 | 135 | ||
| AK392193.1 | - | 2 - 3 | 170 | 135 | ||
| CJ013638.1 | - | 2 - 3 | 422 | 135 | ||
| EW433196.2 | - | 2 - 3 | 211 | 135 | ||
Set Membership: |
|