Search
Search
| Gene Symbol: RBMX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244096.1 | NP_001231025.1 | 6 - 7 | 792 | 91 | |
| XM_013986455.2 | XP_013841909.1 | 847 | 91 | |||
| XM_021079411.1 | XP_020935070.1 | 864 | 91 | |||
| XM_021079412.1 | XP_020935071.1 | 847 | 91 | |||
| GenBank mRNA | AK235135.1 | - | 6 - 7 | 792 | 91 | |
| AK236383.1 | - | 6 - 7 | 811 | 91 | ||
| AK236451.1 | - | 6 - 7 | 795 | 91 | ||
| AK236585.1 | - | 6 - 7 | 796 | 91 | ||
| AK394814.1 | - | 6 - 7 | 792 | 91 | ||
| AK397739.1 | - | 6 - 7 | 847 | 91 | ||
Set Membership: |
|