Search
Search
| Gene Symbol: RPS10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244106.1 | NP_001231035.1 | 4 - 5 | 789 | 65 | |
| XM_005665896.3 | XP_005665953.2 | 541 | 65 | |||
| GenBank mRNA | AK231549.1 | - | 4 - 5 | 436 | 65 | |
| AK240249.1 | - | 4 - 5 | 718 | 65 | ||
| AK349815.1 | - | 4 - 5 | 437 | 65 | ||
| AK349976.1 | - | 4 - 5 | 437 | 65 | ||
| AK350605.1 | - | 4 - 5 | 722 | 65 | ||
| AK397539.1 | - | 4 - 5 | 436 | 65 | ||
Set Membership: |
|