Search
Search
| Gene Symbol: HMGN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244112.1 | NP_001231041.1 | 2 - 3 | 262 | 103 | |
| NM_001244114.1 | NP_001231043.1 | 2 - 3 | 154 | 103 | ||
| GenBank mRNA | AK236192.1 | - | 2 - 3 | 245 | 103 | |
| AK347742.1 | - | 2 - 3 | 192 | 103 | ||
| AK349259.1 | - | 2 - 3 | 252 | 103 | ||
| AK350129.1 | - | 2 - 3 | 460 | 103 | ||
| AK391733.1 | - | 2 - 3 | 159 | 103 | ||
| AK397701.1 | - | 2 - 3 | 252 | 103 | ||
| BW968439.1 | - | 2 - 3 | 262 | 103 | ||
| EW418018.2 | - | 2 - 3 | 154 | 103 | ||
Set Membership: |
|