Search
Search
| Gene Symbol: PSMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244151.1 | NP_001231080.1 | 6 - 7 | 638 | 87 | |
| XM_021098047.1 | XP_020953706.1 | 609 | 87 | |||
| GenBank mRNA | AK235262.1 | - | 6 - 7 | 629 | 87 | |
| AK238210.1 | - | 5 - 6 | 603 | 87 | ||
| AK345545.1 | - | 4 - 5 | 522 | 87 | ||
| AK389202.1 | - | 6 - 7 | 634 | 87 | ||
| AK391703.1 | - | 6 - 7 | 612 | 87 | ||
| AY609681.1 | - | 6 - 7 | 616 | 87 | ||
| FD588173.1 | - | 6 - 7 | 647 | 87 | ||
Set Membership: |
|