Search
Search
| Gene Symbol: SORD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244162.1 | NP_001231091.1 | 2 - 3 | 250 | 77 | |
| GenBank mRNA | AK236025.1 | - | 2 - 3 | 210 | 77 | |
| AK389436.1 | - | 2 - 3 | 250 | 77 | ||
| AK392246.1 | - | 2 - 3 | 238 | 77 | ||
| AY610034.1 | - | 2 - 3 | 165 | 77 | ||
Set Membership: |
|