Search
Search
| Gene Symbol: ZXDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244222.1 | NP_001231151.1 | 6 - 7 | 2126 | 88 | |
| XM_013996783.2 | XP_013852237.1 | 2187 | 88 | |||
| GenBank mRNA | AK347084.1 | - | 1 - 2 | 383 | 88 | |
| BI339313.1 | - | 1 - 2 | 499 | 88 | ||
| EW077240.2 | - | 1 - 2 | 259 | 88 | ||
Set Membership: |
|