Search
Search

CATSPER4| Gene Symbol: CATSPER4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244280.2 | NP_001231209.1 | 3 - 4 | 335 | 76 | |
| XM_021093374.1 | XP_020949033.1 | 76 | ||||
| GenBank mRNA | HQ654522.1 | - | 3 - 4 | 336 | 76 | |
Set Membership: |
|