Search
Search
| Gene Symbol: PRDX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244474.1 | NP_001231403.1 | 2 - 3 | 215 | 94 | |
| GenBank mRNA | AK349436.1 | - | 2 - 3 | 215 | 94 | |
| AK390553.1 | - | 2 - 3 | 164 | 94 | ||
| AK392270.1 | - | 2 - 3 | 183 | 94 | ||
| AK400020.1 | - | 2 - 3 | 215 | 94 | ||
| AY610343.1 | - | 2 - 3 | 151 | 94 | ||
| GQ913659.1 | - | 2 - 3 | 169 | 94 | ||
Set Membership: |
|