Search
Search
| Gene Symbol: AP3S2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244548.1 | NP_001231477.1 | 5 - 6 | 536 | 62 | |
| GenBank mRNA | AK233495.1 | - | 5 - 6 | 497 | 62 | |
| AK391903.1 | - | 5 - 6 | 537 | 62 | ||
| AK399059.1 | - | 3 - 4 | 230 | 62 | ||
| AY609451.1 | - | 5 - 6 | 496 | 62 | ||
| BW976052.1 | - | 5 - 6 | 497 | 62 | ||
Set Membership: |
|