Search
Search

TRAPPC13| Gene Symbol: TRAPPC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244746.1 | NP_001231675.1 | 3 - 4 | 512 | 96 | |
| XM_005672485.3 | XP_005672542.1 | 535 | 96 | |||
| XM_005672486.3 | XP_005672543.1 | 535 | 96 | |||
| XM_005672487.3 | XP_005672544.1 | 572 | 96 | |||
| GenBank mRNA | AK231023.1 | - | 3 - 4 | 512 | 96 | |
| AY609902.1 | - | 3 - 4 | 513 | 96 | ||
Set Membership: |
|