Search
Search
| Gene Symbol: CASZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095326.1 | XP_020950985.1 | 3403 | 60 | ||
| XM_021095327.1 | XP_020950986.1 | 3100 | 60 | |||
| XM_021095328.1 | XP_020950987.1 | 3343 | 60 | |||
| XM_021095329.1 | XP_020950988.1 | 3322 | 60 | |||
| XM_021095330.1 | XP_020950989.1 | 3403 | 60 | |||
| XM_021095331.1 | XP_020950990.1 | 3094 | 60 | |||
Set Membership: |
|