Search
Search
| Gene Symbol: UBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021097089.1 | XP_020952748.1 | 2241 | 64 | ||
| XM_021097091.1 | XP_020952750.1 | 3046 | 64 | |||
| XM_021097096.1 | XP_020952755.1 | 1970 | 64 | |||
| XM_021097103.1 | XP_020952762.1 | 1911 | 64 | |||
Set Membership: |
|