Search
Search
| Gene Symbol: TBPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013993442.2 | XP_013848896.1 | 921 | 100 | ||
| XM_021102336.1 | XP_020957995.1 | 1023 | 100 | |||
| XM_021102340.1 | XP_020957999.1 | 1231 | 100 | |||
| XM_021102365.1 | XP_020958024.1 | 1128 | 100 | |||
Set Membership: |
|