Search
Search
| Gene Symbol: LAMC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021063797.1 | XP_020919456.1 | 7171 | 83 | ||
| XM_021063798.1 | XP_020919457.1 | 5513 | 83 | |||
| XM_021063799.1 | XP_020919458.1 | 5554 | 83 | |||
| XM_021063800.1 | XP_020919459.1 | 6207 | 83 | |||
| XM_021063801.1 | XP_020919460.1 | 5438 | 83 | |||
| XM_021063802.1 | XP_020919461.1 | 5255 | 83 | |||
| XM_021063803.1 | XP_020919462.1 | 3429 | 83 | |||
Set Membership: |
|