Search
Search
| Gene Symbol: EYA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021077393.1 | XP_020933052.1 | 504 | 66 | ||
| XM_021077394.1 | XP_020933053.1 | 772 | 66 | |||
| XM_021077395.1 | XP_020933054.1 | 456 | 66 | |||
| XM_021077396.1 | XP_020933055.1 | 564 | 66 | |||
| XM_021077397.1 | XP_020933056.1 | 1648 | 66 | |||
| XM_021077398.1 | XP_020933057.1 | 494 | 66 | |||
| XM_021077399.1 | XP_020933058.1 | 601 | 66 | |||
Set Membership: |
|