Search
Search
| Gene Symbol: NLRP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095065.1 | XP_020950724.1 | 2307 | 78 | ||
| XM_021095066.1 | XP_020950725.1 | 2362 | 78 | |||
| XM_021095067.1 | XP_020950726.1 | 2329 | 78 | |||
| XM_021095068.1 | XP_020950727.1 | 2307 | 78 | |||