Search
Search

DPY19L2| Gene Symbol: DPY19L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134795.4 | XP_003134843.2 | 1220 | 90 | ||
| XM_005657778.3 | XP_005657835.2 | 1220 | 90 | |||
| XM_021078556.1 | XP_020934215.1 | 1220 | 90 | |||
| XM_021078557.1 | XP_020934216.1 | 1220 | 90 | |||
| XM_021078558.1 | XP_020934217.1 | 1220 | 90 | |||
| XM_021078559.1 | XP_020934218.1 | 1220 | 90 | |||