Search
Search
| Gene Symbol: EXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005659737.3 | XP_005659794.1 | 891 | 71 | ||
| XM_005659741.3 | XP_005659798.1 | 685 | 71 | |||
| XM_013993164.2 | XP_013848618.1 | 774 | 71 | |||
| XM_013993165.2 | XP_013848619.1 | 1968 | 71 | |||
| XM_013993166.2 | XP_013848620.1 | 911 | 71 | |||
| XM_021097503.1 | XP_020953162.1 | 980 | 71 | |||
| XM_021097534.1 | XP_020953193.1 | 980 | 71 | |||
Set Membership: |
|