Search
Search
| Gene Symbol: USP47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083314.1 | XP_020938973.1 | 905 | 96 | ||
| XM_021083315.1 | XP_020938974.1 | 1123 | 96 | |||
| XM_021083316.1 | XP_020938975.1 | 1071 | 96 | |||
Set Membership: |
|